Learn More
Medchemexpress LLC HY-100727 5mg Medchemexpress, AM-2099 CAS:1443373-17-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007275MG
Encompass_Preferred
Medchemexpress, HY-100727 5mg AM-2099 CAS:1443373-17-8 AM-2099 is a potent and selective inhibitor of voltage-gated sodium channel Nav1.7 with an IC 50 of 0.16 μM for human Nav1.7. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1927