Learn More
Medchemexpress LLC HY-100221 5mg Medchemexpress, AM-2394 CAS:1442684-77-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1002215MG
Encompass_Preferred
Medchemexpress, HY-100221 5mg AM-2394 CAS:1442684-77-6 AM-2394 is a structurally distinct glucokinase activator (GKA). AM-2394 activates glucokinase (GK) with an EC 50 of 60 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1759