Learn More
Enzo Life Sciences AM-404 (50mg). CAS: 183718-77-6
SDP
Supplier:  Enzo Life Sciences BMLCR1060050
Encompass_Preferred
Analog of anandamide (BML-FA017). Potentiates the activity of endogenous anandamide by blocking its re-uptake into presynaptic membranes. Activates TRPV1 (EC50=0.04µM) at concentrations lower than those required to inhibit anandamide transport into the cell (neuronal (C6 glioma cells): IC50=10µM; non-neuronal (rat RBL-2H3 cells): IC50=11µM). Low affinity to FAAH (IC50=5.9µM), to CB1 receptor (IC50=1.76µM) and to CB2 receptor (IC50>1µM). Alternative name: N-(4-Hydroxyphenyl)-5Z 8Z 11Z 14Z-eicosatetraenamide, N-(4-Hydroxyphenyl)arachidonoylamide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0470