Learn More
Medchemexpress LLC AM-5308 | 2410796-89-1 | 99.2% | C26H35N5O5S | 50 MG
SDP
Catalog No. 5000355066
Encompass_Preferred
AM-5308 is an inhibitor for KIF18A that inhibits KIF18A-mediated microtubule ATPase activity. It activates mitotic checkpoints, regulates cell division processes, and exhibits antitumor activity.
  • Inhibits KIF18A-mediated microtubule ATPase activity
  • Activates mitotic checkpoints
  • Regulates cell division processes, including chromosome segregation and spindle assembly
  • Exhibits antitumor activity
  • Shows cytotoxicity in human bone marrow cells in vitro
  • Arrests cell cycle of MDA-MB-157 and human bone marrow cells in mitosis phase in vitro
  • Demonstrates antitumor activity in mouse OVCAR-3 xenograft models in vivo
Catalog No. 50-003-55066 Supplier Medchemexpress LLC Supplier No. HY14489450MG
AM-5308 is an inhibitor for KIF18A that inhibits KIF18A-mediated microtubule ATPase activity. It activates mitotic checkpoints, regulates cell division processes, and exhibits antitumor activity.
  • Inhibits KIF18A-mediated microtubule ATPase activity
  • Activates mitotic checkpoints
  • Regulates cell division processes, including chromosome segregation and spindle assembly
  • Exhibits antitumor activity
  • Shows cytotoxicity in human bone marrow cells in vitro
  • Arrests cell cycle of MDA-MB-157 and human bone marrow cells in mitosis phase in vitro
  • Demonstrates antitumor activity in mouse OVCAR-3 xenograft models in vivo