Learn More
Enzo Life Sciences AM-580 (5mg). CAS: 102121-60-8
SDP
Supplier:  Enzo Life Sciences BMLGR1040005
Encompass_Preferred
Retinoic acid analog which acts as a selective RARα agonist with no affinity for RXRs. Induces differentiation in leukemia cell lines and inhibits angiogenesis in chick embryos. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) and ethanol (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0916