Learn More
Enzo Life Sciences AM-630 (10mg). CAS: 164178-33-0
SDP
Catalog No. 502010471
Encompass_Preferred
Acts as a cannabinoid receptor antagonist in mouse brain, vas deferens and guinea pig brain, but an agonist in guinea pig ileum. Acts as an inverse agonist on cloned human CB1 receptors. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-201-0471 Supplier Enzo Life Sciences Supplier No. BMLCR1070010
Acts as a cannabinoid receptor antagonist in mouse brain, vas deferens and guinea pig brain, but an agonist in guinea pig ileum. Acts as an inverse agonist on cloned human CB1 receptors. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: -20°C.