Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AM-6538 | 1245626-00-9 | 99.9% | 5 MG
SDP

Catalog No. 5000234116
Encompass_Preferred

AM-6538 is a small-molecule cannabinoid receptor antagonist supplied for preclinical research. It is provided as a white to off-white powder and is characterized by LCMS analysis and a certificate of analysis indicating high purity.

  • Small-molecule cannabinoid receptor antagonist for preclinical research.
  • White to off-white powder with LCMS-confirmed identity.
  • High purity (LCMS): 99.85%.
  • Molecular formula C26H25Cl2N5O4; molecular weight 542.41 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.
  • Supplied in small pack sizes suitable for assay development and biochemical studies.

Catalog No. 50-002-34116 Supplier Medchemexpress LLC Supplier No. HY1204235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

AM-6538 is a small-molecule cannabinoid receptor antagonist supplied for preclinical research. It is provided as a white to off-white powder and is characterized by LCMS analysis and a certificate of analysis indicating high purity.

  • Small-molecule cannabinoid receptor antagonist for preclinical research.
  • White to off-white powder with LCMS-confirmed identity.
  • High purity (LCMS): 99.85%.
  • Molecular formula C26H25Cl2N5O4; molecular weight 542.41 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.
  • Supplied in small pack sizes suitable for assay development and biochemical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.