Learn More
Medchemexpress LLC AM-92016 hydrochloride | 133229-11-5 | 99.3% | 483.84 g/mol | C19H25Cl3N2O4S | 10 MG
SDP
Catalog No. 5000211113
Encompass_Preferred
AM-92016 hydrochloride is a research-grade small molecule that selectively blocks the rectifier potassium current (IK), delaying IK-mediated repolarization and restricting action potential duration. It is supplied as a high-purity solid intended for use in electrophysiology and ion-channel research.
  • Specific blocker of rectifier potassium current (IK).
  • Suitable for electrophysiology and ion-channel research.
  • High purity (99.3%) for reliable experimental results.
  • Solid, light yellow to yellow appearance.
  • Storage: sealed at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size 10 mg for small-scale studies.
Catalog No. 50-002-11113 Supplier Medchemexpress LLC Supplier No. HY10125310MG
AM-92016 hydrochloride is a research-grade small molecule that selectively blocks the rectifier potassium current (IK), delaying IK-mediated repolarization and restricting action potential duration. It is supplied as a high-purity solid intended for use in electrophysiology and ion-channel research.
  • Specific blocker of rectifier potassium current (IK).
  • Suitable for electrophysiology and ion-channel research.
  • High purity (99.3%) for reliable experimental results.
  • Solid, light yellow to yellow appearance.
  • Storage: sealed at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size 10 mg for small-scale studies.