Learn More
Medchemexpress LLC AM-92016 hydrochloride | 133229-11-5 | MFCD00673947 | 99.3% | 483.84 | C19H25Cl3N2O4S | 1 ML
SDP
Catalog No. 5000211706
Encompass_Preferred
AM-92016 hydrochloride is a small-molecule blocker of delayed rectifier potassium currents (IK), commonly used in electrophysiology and cardiovascular research to modulate action potential duration. It is supplied both as a solid and as a ready-to-use solution to support in vitro pharmacology and ion-channel assays.
  • Blocks delayed rectifier potassium channels (IK) and shortens action potential duration.
  • Supplied as a 10 mM solution in DMSO or as a dry solid for custom preparation.
  • High purity suitable for research use (reported ~99.3%).
  • Molecular weight 483.84 and CAS 133229-11-5 for clear identification.
  • Available in small-quantity formats compatible with pharmacological assays.
Catalog No. 50-002-11706 Supplier Medchemexpress LLC Supplier No. HY10125310MM 1ML
AM-92016 hydrochloride is a small-molecule blocker of delayed rectifier potassium currents (IK), commonly used in electrophysiology and cardiovascular research to modulate action potential duration. It is supplied both as a solid and as a ready-to-use solution to support in vitro pharmacology and ion-channel assays.
  • Blocks delayed rectifier potassium channels (IK) and shortens action potential duration.
  • Supplied as a 10 mM solution in DMSO or as a dry solid for custom preparation.
  • High purity suitable for research use (reported ~99.3%).
  • Molecular weight 483.84 and CAS 133229-11-5 for clear identification.
  • Available in small-quantity formats compatible with pharmacological assays.