Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AM 92016 hydrochloride, Tocris Bioscience™

Catalog No. 087610
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-761-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-761-0 Supplier R&D Systems Supplier No. 0876/10
This item is not returnable. View return policy

KV channel blocker

A specific blocker of the time dependent delayed rectifier potassium current, devoid of any β-adrenoceptor blocking activity. Exhibits proarrhythmic and prohypertensive activity in vivo.

Chemical Identifiers

CAS 133229-11-5
Molecular Formula C19H25Cl3N2O4S
Molecular Weight (g/mol) 483.829
InChI Key TXOARFPCQOBODS-UHFFFAOYSA-N
Synonym am 92016 hydrochloride, 1-4-methanesulfonamidophenoxy-3-n-methyl-3,4-dichlorophenylethylamino-2-propanol hydrochloride, n-4-3-2-3,4-dichlorophenyl ethyl methyl amino-2-hydroxypropoxy phenyl methanesulfonamide hydrochloride
PubChem CID 45073415
IUPAC Name N-[4-[3-[2-(3,4-dichlorophenyl)ethyl-methylamino]-2-hydroxypropoxy]phenyl]methanesulfonamide;hydrochloride
SMILES CN(CCC1=CC(=C(C=C1)Cl)Cl)CC(COC2=CC=C(C=C2)NS(=O)(=O)C)O.Cl

Specifications

Quantity 10 mg
Formula Weight 483.84
Percent Purity >98%
Chemical Name or Material AM 92016 hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.