Learn More
Medchemexpress LLC AM-9747 | 2691869-82-4 | 99.76% | 466.46 | 250 MG
SDP
Catalog No. 5000363255
Encompass_Preferred
AM-9747 (compound 503) is a potent PRMT5 inhibitor with a Ki value of 0.06 nM. It shows antiproliferative activity against HCT116-MTAP null cells (IC50 0.026 μM) and HCT116-WT cells (IC50 0.068 μM) after 6 days.
  • Potent PRMT5 inhibitor
  • Antiproliferative activity
  • High purity of 99.76%
Catalog No. 50-003-63255 Supplier Medchemexpress LLC Supplier No. 250MGHY148674
AM-9747 (compound 503) is a potent PRMT5 inhibitor with a Ki value of 0.06 nM. It shows antiproliferative activity against HCT116-MTAP null cells (IC50 0.026 μM) and HCT116-WT cells (IC50 0.068 μM) after 6 days.
  • Potent PRMT5 inhibitor
  • Antiproliferative activity
  • High purity of 99.76%