Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AM-Imidazole-PA-Boc | 2357108-99-5 | 98.2% | 25 MG
SDP

Catalog No. 5000341590
Encompass_Preferred

AM-Imidazole-PA-Boc is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTAC IRAK4 degrader-1. This product is for research use only and not intended for sale to patients.

  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTAC IRAK4 degrader-1
  • For research use only
  • Not intended for sale to patients
  • CAS number: 2357108-99-5
  • Molecular weight: 254.33
  • Formula: C12H22N4O2
  • Appearance: Oil
  • Color: Off-white to gray
  • Purity: 98.22%

Catalog No. 50-003-41590 Supplier Medchemexpress LLC Supplier No. 25MGHY129968
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AM-Imidazole-PA-Boc is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTAC IRAK4 degrader-1. This product is for research use only and not intended for sale to patients.

  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTAC IRAK4 degrader-1
  • For research use only
  • Not intended for sale to patients
  • CAS number: 2357108-99-5
  • Molecular weight: 254.33
  • Formula: C12H22N4O2
  • Appearance: Oil
  • Color: Off-white to gray
  • Purity: 98.22%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.