Learn More
Medchemexpress LLC AM-Imidazole-PA-Boc | 2357108-99-5 | 98.2% | 250 MG
SDP
Catalog No. 5000341533
Encompass_Preferred
AM-Imidazole-PA-Boc is an alkyl chain-based PROTAC linker. This compound is intended for research applications, particularly in the synthesis of PROTAC IRAK4 degrader-1. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system, facilitating the selective degradation of target proteins. They are characterized by two distinct ligands, linked together, where one ligand binds to an E3 ubiquitin ligase and the other to the target protein.
  • Alkyl chain-based PROTAC linker
  • Can be used in the synthesis of PROTAC IRAK4 degrader-1
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Contains two different ligands connected by a linker
Catalog No. 50-003-41533 Supplier Medchemexpress LLC Supplier No. 250MGHY129968
AM-Imidazole-PA-Boc is an alkyl chain-based PROTAC linker. This compound is intended for research applications, particularly in the synthesis of PROTAC IRAK4 degrader-1. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system, facilitating the selective degradation of target proteins. They are characterized by two distinct ligands, linked together, where one ligand binds to an E3 ubiquitin ligase and the other to the target protein.
  • Alkyl chain-based PROTAC linker
  • Can be used in the synthesis of PROTAC IRAK4 degrader-1
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Contains two different ligands connected by a linker