Learn More
Medchemexpress LLC AM-Imidazole-PA-Boc | 2357108-99-5 | 98.22% | 1 G
SDP
Catalog No. 5000340078
Encompass_Preferred
AM-Imidazole-PA-Boc is an alkyl chain-based PROTAC linker primarily used in the synthesis of PROTAC IRAK4 degrader-1 (HY-129966). It is intended for research use only and not for medical or diagnostic applications.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTAC IRAK4 degrader-1
  • For research use only
Catalog No. 50-003-40078 Supplier Medchemexpress LLC Supplier No. HY1299681G
AM-Imidazole-PA-Boc is an alkyl chain-based PROTAC linker primarily used in the synthesis of PROTAC IRAK4 degrader-1 (HY-129966). It is intended for research use only and not for medical or diagnostic applications.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTAC IRAK4 degrader-1
  • For research use only