Learn More
Medchemexpress LLC AM095 | 1345614-59-6 | 99.7% | 478.47 | 200 MG
SDP
Catalog No. 5000394402
Encompass_Preferred
AM095 is a selective LPA1 receptor antagonist known for its potent inhibition of LPA-induced calcium flux in human and mouse LPA1-transfected CHO cells. It demonstrates an exceedingly low IC50 for human (0.025 μM) and mouse (0.023 μM) cells, positioning it as a valuable tool for research involving LPA1 receptor pathways.
  • Selective LPA1 receptor antagonist.
  • Potently inhibits LPA-induced calcium flux.
  • Applicable in studies involving human or mouse LPA1-transfected CHO cells.
  • Useful for researching LPA1 receptor pathways.
Catalog No. 50-003-94402 Supplier Medchemexpress LLC Supplier No. HY16039200MG
AM095 is a selective LPA1 receptor antagonist known for its potent inhibition of LPA-induced calcium flux in human and mouse LPA1-transfected CHO cells. It demonstrates an exceedingly low IC50 for human (0.025 μM) and mouse (0.023 μM) cells, positioning it as a valuable tool for research involving LPA1 receptor pathways.
  • Selective LPA1 receptor antagonist.
  • Potently inhibits LPA-induced calcium flux.
  • Applicable in studies involving human or mouse LPA1-transfected CHO cells.
  • Useful for researching LPA1 receptor pathways.