Learn More
Apexbio Technology LLC AM095 1345614-59-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064096
Encompass_Preferred
AM095 (CAS 1345614-59-6) is an orally bioavailable antagonist targeting the lysophosphatidic acid receptor 1 (LPA1) It selectively inhibits recombinant human and mouse LPA1 receptors with IC50 values of 0 98 M and 0 73 M respectively In cell-based assays AM095 suppresses LPA1-mediated chemotaxis in mouse LPA1/CHO and human melanoma A2058 cells (IC50 values 0 78 M and 0 23 M respectively) In vivo studies demonstrate its ability to prevent LPA-triggered histamine release reduce collagen and protein accumulation and limit inflammatory cell infiltration in mouse pulmonary models AM095 serves as a valuable tool for investigating LPA1-associated pathologies
Catalog No. 50-000-64096 Supplier Apexbio Technology LLC Supplier No. A31665.1
 Restricted Item
Add to Cart
Add to Cart
AM095 (CAS 1345614-59-6) is an orally bioavailable antagonist targeting the lysophosphatidic acid receptor 1 (LPA1) It selectively inhibits recombinant human and mouse LPA1 receptors with IC50 values of 0 98 M and 0 73 M respectively In cell-based assays AM095 suppresses LPA1-mediated chemotaxis in mouse LPA1/CHO and human melanoma A2058 cells (IC50 values 0 78 M and 0 23 M respectively) In vivo studies demonstrate its ability to prevent LPA-triggered histamine release reduce collagen and protein accumulation and limit inflammatory cell infiltration in mouse pulmonary models AM095 serves as a valuable tool for investigating LPA1-associated pathologies