Learn More
Medchemexpress LLC AM095 free acid | 1228690-36-5 | 99.0% | 456.49 | 200 MG
SDP
Catalog No. 5000391678
Encompass_Preferred
AM095 free acid is a potent LPA1 receptor antagonist that effectively inhibits LPA-induced calcium flux in CHO cells transfected with human or mouse LPA1. It also significantly reduces LPA-induced vasorelaxation and inhibits LPA-driven chemotaxis in various cell types. In preclinical studies, it has shown to attenuate bleomycin-induced dermal fibrosis and decrease kidney fibrosis in a mouse model of unilateral ureteral obstruction. The compound exhibits high oral bioavailability, a moderate half-life, and is well tolerated in animal models.
  • Potent LPA1 receptor antagonist.
  • Inhibits LPA-induced calcium flux in CHO cells.
  • Reduces LPA-induced vasorelaxation.
  • Inhibits LPA-driven chemotaxis in melanoma cells.
  • Attenuates bleomycin-induced dermal fibrosis in vivo.
  • Decreases kidney fibrosis in a mouse model.
  • High oral bioavailability.
  • Moderate half-life.
  • Well tolerated in rats and dogs.
Catalog No. 50-003-91678 Supplier Medchemexpress LLC Supplier No. HY16040200MG
AM095 free acid is a potent LPA1 receptor antagonist that effectively inhibits LPA-induced calcium flux in CHO cells transfected with human or mouse LPA1. It also significantly reduces LPA-induced vasorelaxation and inhibits LPA-driven chemotaxis in various cell types. In preclinical studies, it has shown to attenuate bleomycin-induced dermal fibrosis and decrease kidney fibrosis in a mouse model of unilateral ureteral obstruction. The compound exhibits high oral bioavailability, a moderate half-life, and is well tolerated in animal models.
  • Potent LPA1 receptor antagonist.
  • Inhibits LPA-induced calcium flux in CHO cells.
  • Reduces LPA-induced vasorelaxation.
  • Inhibits LPA-driven chemotaxis in melanoma cells.
  • Attenuates bleomycin-induced dermal fibrosis in vivo.
  • Decreases kidney fibrosis in a mouse model.
  • High oral bioavailability.
  • Moderate half-life.
  • Well tolerated in rats and dogs.