Learn More
Biotium AM1-43
Catalog No. NC1669431
Encompass_Preferred
AM1-43 is a fixable, activity-dependent fluorescent nerve terminal probe. AM1-43 functions similarly similarly to SynaptoGreen™ C4, but with the additional property of being fixable. Thus, AM1-43 is a useful tool for synapse studies where subsequent fluorescent immunochemistry is desired. AM1-43 and FM1-43 have identical excitation and emission wavelengths. Also see AM1-44, which is similar to AM1-43, but has longer lipophilic tails, resulting in better fixability.Features: Ex/Em nm: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C29H49Cl3N4; MW: 559.5
Catalog No. NC1669431 Supplier Biotium Supplier No. 70024
AM1-43 is a fixable, activity-dependent fluorescent nerve terminal probe. AM1-43 functions similarly similarly to SynaptoGreen™ C4, but with the additional property of being fixable. Thus, AM1-43 is a useful tool for synapse studies where subsequent fluorescent immunochemistry is desired. AM1-43 and FM1-43 have identical excitation and emission wavelengths. Also see AM1-44, which is similar to AM1-43, but has longer lipophilic tails, resulting in better fixability.Features: Ex/Em nm: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C29H49Cl3N4; MW: 559.5