Learn More
Biotium AM1-44
Supplier:  Biotium 70038
Encompass_Preferred
AM1-44 is a version of AM1-43 with improved fixability. The dye can also be used as a general probe for following endocytosis. AM1-44 probe functions similarly as AM1-43, except that AM1-44 has lipophilic tails that are each one carbon longer than those of AM1-43, resulting in better fixability. Properties: Ex/Em: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C31H52Cl3N4; MW: 587.5
Catalog No. NC1669432