Learn More
Cayman Chemical AM1220 azepn Isomer 5mg
SDP
Catalog No. 502952615
Encompass_Preferred
An isomer of AM1220 in which the piperidine group has been replaced with azepane; intended for forensic and research applications
Catalog No. 50-295-2615 Supplier Cayman Chemical Supplier No. 115835 MG
An isomer of AM1220 in which the piperidine group has been replaced with azepane; intended for forensic and research applications