Learn More
TARGETMOL CHEMICALS INC AM1241 25MG
SDP
Catalog No. 502579291
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AM-1241 is a selective cannabinoid CB2 receptor agonist with Ki of 3.4 nM exhibits 82-fold selectivity over CB1 receptor. purity: 98%
Catalog No. 50-257-9291 Supplier TARGETMOL CHEMICALS INC Supplier No. T675525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AM-1241 is a selective cannabinoid CB2 receptor agonist with Ki of 3.4 nM exhibits 82-fold selectivity over CB1 receptor. purity: 98%