Learn More
Selleck Chemical LLC AM1241 S1544-2mg
SDP
Catalog No. 503108470
Encompass_Preferred
AM-1241 is a selective cannabinoid CB2 receptor agonist with Ki of 3 4 nM exhibits 82-fold selectivity over CB1 receptor
Catalog No. 50-310-8470 Supplier Selleck Chemical LLC Supplier No. S15442MG
AM-1241 is a selective cannabinoid CB2 receptor agonist with Ki of 3 4 nM exhibits 82-fold selectivity over CB1 receptor