Learn More
Cayman Chemical AM2201 2-naphthyl Isomer 1mg
SDP
Catalog No. 502951103
Encompass_Preferred
Differs structurally from AM2201 by having the naphthyl group attached at the 2' position
Catalog No. 50-295-1103 Supplier Cayman Chemical Supplier No. 108621 MG
Differs structurally from AM2201 by having the naphthyl group attached at the 2' position