Learn More
Apexbio Technology LLC AM251, 10mg. Cas: 183232-66-8 MFCD: MFCD01861181. Potent CB1 antagonist
SDP
Catalog No. 501012168
Encompass_Preferred
AM251 block the inhibitory effects of endocannabinoids and synthetic cannabinoid agonists on transmitter release through an action at presynaptic cannabinoid 1 receptors in brain. 183232-66-8. MFCD01861181
Catalog No. 50-101-2168 Supplier Apexbio Technology LLC Supplier No. B142710
AM251 block the inhibitory effects of endocannabinoids and synthetic cannabinoid agonists on transmitter release through an action at presynaptic cannabinoid 1 receptors in brain. 183232-66-8. MFCD01861181