Learn More
Selleck Chemical LLC AM251-5mg
SDP
Catalog No. 502389940
Encompass_Preferred
AM251 block the inhibitory effects of endocannabinoids and synthetic cannabinoid agonists on transmitter release through an action at presynaptic cannabinoid 1 receptors in brain.
Catalog No. 50-238-9940 Supplier Selleck Chemical LLC Supplier No. S28195MG
AM251 block the inhibitory effects of endocannabinoids and synthetic cannabinoid agonists on transmitter release through an action at presynaptic cannabinoid 1 receptors in brain.