Learn More
Medchemexpress LLC HY-15443 5mg Medchemexpress, AM251 CAS:183232-66-8 Purity:>98%
SDP
Catalog No. 502081724
Encompass_Preferred
Medchemexpress, HY-15443 5mg AM251 CAS:183232-66-8 AM251 is a selective cannabinoid 1 (CB1) receptor antagonist with an IC50 of 8 nM, also acts as a potent GPR55 agonist with an EC50 of 39 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1724 Supplier Medchemexpress LLC Supplier No. HY154435MG
Medchemexpress, HY-15443 5mg AM251 CAS:183232-66-8 AM251 is a selective cannabinoid 1 (CB1) receptor antagonist with an IC50 of 8 nM, also acts as a potent GPR55 agonist with an EC50 of 39 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.