Learn More
WELCH ALLYN INC Welch Allyn AM282 Manual Audiometer with 11 Frequencies (125 Hz to 8,000 Hz)
Catalog No. NC1388191
Encompass
Welch Allyn AM282 Manual Audiometer with 11 Frequencies (125 Hz to 8,000 Hz), Audiometry Headset included, Threshold Audiometry Card, Audiogram Forms (Qty. 50), Soft Storage Case; 100-240 V, 50-60 Hz AC, IEC Plug Type-B
This item is not returnable. View return policy
Catalog No. NC1388191 Supplier WELCH ALLYN INC Supplier No. 28200
Welch Allyn AM282 Manual Audiometer with 11 Frequencies (125 Hz to 8,000 Hz), Audiometry Headset included, Threshold Audiometry Card, Audiogram Forms (Qty. 50), Soft Storage Case; 100-240 V, 50-60 Hz AC, IEC Plug Type-B