Learn More
Apexbio Technology LLC AM580, 102121-60-8, MFCD00673916, 10 mM in 1 mL DMSO.
SDP
Supplier:  Apexbio Technology LLC B46545.1
Encompass_Preferred
MF: C22H25NO3, MW: 351.44, Solubility: 14.3mg/mL in DMSO. AM580 is a selective agonist of retinoic acid receptor (RAR) with IC50 and EC50 values of 8 nM and 0.36 nM, respectively.
Catalog No. 50-115-1828