Learn More
TARGETMOL CHEMICALS INC AM580 25MG
SDP
Catalog No. 502579727
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AM580 (CD336) is a retinoic acid receptor agonist that is selective for RAR(alpha)( IC50 8 nM )purity: 99%
Catalog No. 50-257-9727 Supplier TARGETMOL CHEMICALS INC Supplier No. T585425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AM580 (CD336) is a retinoic acid receptor agonist that is selective for RAR(alpha)( IC50 8 nM )purity: 99%