Learn More
Medchemexpress LLC HY-10475 5mg Medchemexpress, AM580 CAS:102121-60-8 Purity:>98%
SDP
Catalog No. 501968231
Encompass_Preferred
Medchemexpress, HY-10475 5mg AM580 CAS:102121-60-8 AM580 is a selective RARα agonist with IC50 and EC50 of 8 nM and 0.36 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8231 Supplier Medchemexpress LLC Supplier No. HY104755MG
Medchemexpress, HY-10475 5mg AM580 CAS:102121-60-8 AM580 is a selective RARα agonist with IC50 and EC50 of 8 nM and 0.36 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.