Learn More
Selleck Chemical LLC AM580-5mg
SDP
Catalog No. 502390005
Encompass_Preferred
AM580 is a retinoic acid receptor agonist with Kd values of 8 nM, 131 nM and 450 nM for RARα, RARβ and RARγ.
Catalog No. 50-239-0005 Supplier Selleck Chemical LLC Supplier No. S29335MG
AM580 is a retinoic acid receptor agonist with Kd values of 8 nM, 131 nM and 450 nM for RARα, RARβ and RARγ.