Learn More
STEMCELL Technologies AM580, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72964
Encompass
Selectively activate retinoic acid receptor alpha (RARα) compared to RARβ and RARγ with AM580 (Bernard et al. Biochem Biophys Res Commun, 1992). Used to reprogram somatic cells to induced pluripotent stem cells (Wang et al. Proc Natl Acad Sci USA, 2011), this retinoid pathway activator also inhibits tumor growth in MMTV-Myc mice (Bosch et al. Breast Cancer Res, 2012); induces differentiation in acute promyelocytic leukemia cells (Gianní et al. Blood, 1996); and inhibits mouse mammary tumor virus (MMTV)-neu- and MMTV-wnt1-induced mammary gland hyperplasia (Lu et al. Oncogene, 2010). AM580 is also used to induce differentiation of human induced pluripotent stem cells into intermediate mesoderm, in combination with the CHIR99021 (Araoka et al. PLoS One, 2014). AM580 is supplied as a crystalline solid with ≥98% purity; Molecular weight 351.4 g/mol; CAS Number 102121-60-8. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-6007
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov