Learn More
Cayman Chemical AM630, 164178-33-0, 10 mg
SDP
Supplier:  Cayman Chemical 1000697410
Encompass_Preferred
Molecular Formula C23H25IN2O3, Purity ≥98%, Formula Weight 504.4, Synonyms: [6-iodo-2-methyl-1-[2-(4-morpholinyl)ethyl]-1H-indol-3-yl](4-methoxyphenyl)-methanone, 6-Iodopravodoline; An inverse agonist at CB2 and partial agonist at CB1 receptors; binds selectively to CB2 over CB1 receptors (Kis = 31.2 and 5,152 nM, respectively); blocks the antinociceptive effects of cannabinoids; decreases proliferation and induces cell cycle arrest in renal cell carcinoma cells.
Catalog No. NC0587960
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.