Learn More
Cayman Chemical AM630 Other Receptor Modulator
Supplier: Cayman Chemical 1000697410 MG
An inverse agonist at CB2 and partial agonist at CB1 receptors; binds selectively to CB2 over CB1 receptors (Kis = 31.2 and 5,152 nM, respectively); blocks the antinociceptive effects of cannabinoids; decreases proliferation and induces cell cycle arrest in renal cell carcinoma cells