Learn More
eMolecules​ 4-[(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl]benzoic acid | Combi-Blocks | 94497-51-5 | MFCD00866188 | 351.446 | C22H25NO3 | 97.000 | CC1(C)CCC(C)(C)c2cc(NC(=O)c3ccc(cc3)C(O)=O)ccc12 | 1g | 228814772
Supplier:  eMolecules​ 228814772
Encompass_Preferred
4-[(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl]benzoic acid | Combi-Blocks | 94497-51-5 | MFCD00866188 | 351.446 | C22H25NO3 | 97.000 | CC1(C)CCC(C)(C)c2cc(NC(=O)c3ccc(cc3)C(O)=O)ccc12 | 1g | 228814772
Catalog No. NC2009807