Learn More
Cayman Chemical AM92016 (hydrochloride), Purity: ≥98%, CAS: 133229-11-5, 10 mg
SDP
Catalog No. 502909668
Encompass_Preferred
An IKr inhibitor (IC50 = 40 nM in isolated rabbit sinoatrial node cells); selective for IKr over L-type Cav current and If at 1 µM; decreases the diastolic depolarization rate and increases cycle length and action potential duration at full repolarization in isolated rabbit sinoatrial node cells at 50 nM; decreases the QT interval and increases heart rate in anesthetized guinea pigs at 1 mg/kg; decreases the time to first arrhythmia and ventricular fibrillation in ouabain-stimulated guinea pigs at 1 mg/kg
Catalog No. 50-290-9668 Supplier Cayman Chemical Supplier No. 3680310 MG
An IKr inhibitor (IC50 = 40 nM in isolated rabbit sinoatrial node cells); selective for IKr over L-type Cav current and If at 1 µM; decreases the diastolic depolarization rate and increases cycle length and action potential duration at full repolarization in isolated rabbit sinoatrial node cells at 50 nM; decreases the QT interval and increases heart rate in anesthetized guinea pigs at 1 mg/kg; decreases the time to first arrhythmia and ventricular fibrillation in ouabain-stimulated guinea pigs at 1 mg/kg