Learn More
Apexbio Technology LLC AM966 1228690-19-4 200mg
SDP
Catalog No. 5000064188
Encompass_Preferred
AM966 (CAS 1228690-19-4) is an orally bioavailable small-molecule antagonist that selectively targets Lysophosphatidic acid type 1 receptor (LPA1) AM966 exhibits potent inhibitory activity against LPA1 receptors from humans and mice with IC50 values of approximately 17 nM and 19 nM respectively demonstrating selectivity over other LPA receptor subtypes (LPA2 5) In vitro AM966 blocks LPA-mediated chemotactic responses in IMR-90 human lung fibroblasts A2058 human melanoma cells and CHO cell lines In vivo AM966 effectively attenuates lung fibrosis inflammation and associated weight loss in bleomycin-induced murine pulmonary fibrosis models highlighting its utility in research on fibrotic diseases
Catalog No. 50-000-64188 Supplier Apexbio Technology LLC Supplier No. A3170200
AM966 (CAS 1228690-19-4) is an orally bioavailable small-molecule antagonist that selectively targets Lysophosphatidic acid type 1 receptor (LPA1) AM966 exhibits potent inhibitory activity against LPA1 receptors from humans and mice with IC50 values of approximately 17 nM and 19 nM respectively demonstrating selectivity over other LPA receptor subtypes (LPA2 5) In vitro AM966 blocks LPA-mediated chemotactic responses in IMR-90 human lung fibroblasts A2058 human melanoma cells and CHO cell lines In vivo AM966 effectively attenuates lung fibrosis inflammation and associated weight loss in bleomycin-induced murine pulmonary fibrosis models highlighting its utility in research on fibrotic diseases