Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 23600MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: AMACR/1723. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: HEK cells or Prostate Adenocarcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length AMACR protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 54kDa.Chromosome: 5p13.3. Gene Symbol: AMACR. Gene ID: 23600. Ensemble: ENSG00000242110. Uniprot: NP_001161067.
Catalog No. 50-188-4706