Learn More
Aviva Systems Biology Corp AMACR Antibody - middle region (ARP60807_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60807_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMACR. It was validated on Western Blot. . Alternate names: CBAS4, RACE, RM, AMACRDApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-172