Learn More
Apexbio Technology LLC Amadacycline 389139-89-3 200mg
SDP
Catalog No. 5000095259
Encompass_Preferred
Amadacycline (CAS 389139-89-3) is a tetracycline-class antibiotic targeting the bacterial ribosomal 30S subunit It is designed to inhibit bacterial protein synthesis thereby preventing microbial proliferation Amadacycline exerts its biological activity primarily through interference with bacterial translation by inhibiting the 30S ribosomal subunit The exact IC50 value for Amadacycline has not been established Based on these pharmacological properties Amadacycline holds research potential in investigating antibiotic resistance mechanisms bacterial ribosome-targeting strategies and as a candidate in studies evaluating therapeutic agents for complicated skin and skin structure infections
Catalog No. 50-000-95259 Supplier Apexbio Technology LLC Supplier No. A3171200
Amadacycline (CAS 389139-89-3) is a tetracycline-class antibiotic targeting the bacterial ribosomal 30S subunit It is designed to inhibit bacterial protein synthesis thereby preventing microbial proliferation Amadacycline exerts its biological activity primarily through interference with bacterial translation by inhibiting the 30S ribosomal subunit The exact IC50 value for Amadacycline has not been established Based on these pharmacological properties Amadacycline holds research potential in investigating antibiotic resistance mechanisms bacterial ribosome-targeting strategies and as a candidate in studies evaluating therapeutic agents for complicated skin and skin structure infections