Learn More
Apexbio Technology LLC β-Amanitin 21150-22-1 1mg
SDP
Catalog No. 5000091361
Encompass_Preferred
beta-Amanitin (CAS 21150-22-1) is a cyclic peptide inhibitor targeting eukaryotic RNA polymerase II and III It modulates transcription processes thereby regulating transcription and subsequent protein synthesis in cells beta-Amanitin exerts its biological activity primarily through inhibition of RNA polymerase II and III Based on these pharmacological properties beta-Amanitin holds research potential in studies of RNA transcriptional regulation cellular response mechanisms to transcriptional stress cell cycle control investigation of transcriptional initiation and elongation analysis of the DNA damage response pathway and elucidation of transcriptional inhibition effects on gene expression in molecular biology and biochemical research
Catalog No. 50-000-91361 Supplier Apexbio Technology LLC Supplier No. B84671
beta-Amanitin (CAS 21150-22-1) is a cyclic peptide inhibitor targeting eukaryotic RNA polymerase II and III It modulates transcription processes thereby regulating transcription and subsequent protein synthesis in cells beta-Amanitin exerts its biological activity primarily through inhibition of RNA polymerase II and III Based on these pharmacological properties beta-Amanitin holds research potential in studies of RNA transcriptional regulation cellular response mechanisms to transcriptional stress cell cycle control investigation of transcriptional initiation and elongation analysis of the DNA damage response pathway and elucidation of transcriptional inhibition effects on gene expression in molecular biology and biochemical research