Learn More
Medchemexpress LLC β-amanitin | 21150-22-1 | MFCD00151212 | >98.0% | 918.96 g/mol | C39H54N10O14S | 1 MG
SDP
Catalog No. 5000203473
Encompass_Preferred
β-Amanitin is a cyclic peptide toxin isolated from Amanita phalloides that potently inhibits eukaryotic RNA polymerase II and, to a lesser extent, RNA polymerase III, blocking mRNA synthesis and protein production. It is used as a biochemical research reagent to study transcription, RNA polymerase function, and as a cytotoxic payload in antibody-drug conjugate research.
  • Potent inhibitor of RNA polymerase II and III.
  • High purity (>98%) suitable for biochemical studies.
  • Soluble in DMSO at high concentration for stock preparation.
  • Appropriate for use as an ADC cytotoxic payload or molecular biology tool.
  • Supplied in small research quantities for laboratory applications.
Catalog No. 50-002-03473 Supplier Medchemexpress LLC Supplier No. HY1255861MG
β-Amanitin is a cyclic peptide toxin isolated from Amanita phalloides that potently inhibits eukaryotic RNA polymerase II and, to a lesser extent, RNA polymerase III, blocking mRNA synthesis and protein production. It is used as a biochemical research reagent to study transcription, RNA polymerase function, and as a cytotoxic payload in antibody-drug conjugate research.
  • Potent inhibitor of RNA polymerase II and III.
  • High purity (>98%) suitable for biochemical studies.
  • Soluble in DMSO at high concentration for stock preparation.
  • Appropriate for use as an ADC cytotoxic payload or molecular biology tool.
  • Supplied in small research quantities for laboratory applications.