Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Amanitin | 23109-05-9 | MFCD00215842 | 99.9% | 918.97 g/mol | C39H54N10O14S | 500 UG
SDP

Catalog No. 5000203729
Encompass_Preferred

α-Amanitin is a bicyclic peptide toxin that potently inhibits RNA polymerase II and is used as a research reagent and ADC payload component. The compound is supplied as a solid; it is unstable in solution and should be stored at -20°C protected from light, with solutions prepared fresh prior to use.

  • Bicyclic peptide inhibitor of RNA polymerase II.
  • High purity (99.9%).
  • CAS number 23109-05-9.
  • Molecular weight 918.97 g/mol.
  • Soluble in water and DMSO with ultrasonic assistance; in vivo formulation protocols are reported.
  • Store at -20°C, protect from light; unstable in solution-prepare fresh solutions.

Catalog No. 50-002-03729 Supplier Medchemexpress LLC Supplier No. HY19610500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Amanitin is a bicyclic peptide toxin that potently inhibits RNA polymerase II and is used as a research reagent and ADC payload component. The compound is supplied as a solid; it is unstable in solution and should be stored at -20°C protected from light, with solutions prepared fresh prior to use.

  • Bicyclic peptide inhibitor of RNA polymerase II.
  • High purity (99.9%).
  • CAS number 23109-05-9.
  • Molecular weight 918.97 g/mol.
  • Soluble in water and DMSO with ultrasonic assistance; in vivo formulation protocols are reported.
  • Store at -20°C, protect from light; unstable in solution-prepare fresh solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.