Learn More
Medchemexpress LLC Amantadine | 768-94-5 | 99.9% | 100 G
SDP
Catalog No. 5000407153
Encompass_Preferred
Amantadine is an orally active and potent antiviral agent effective against influenza A viruses and coronaviruses. It inhibits various ion channels, including NMDA and M2, and shows anti-orthopoxvirus and anticancer activities. This compound is suitable for research applications related to Parkinson's disease, postoperative cognitive dysfunction, and COVID-19.
  • Inhibits influenza A viruses and coronavirus ion channels
  • Demonstrates anti-orthopoxvirus and anticancer activity
  • Suitable for research on Parkinson's disease and COVID-19
  • Inhibits SARS-CoV-2 replication in Vero E6 cells
  • Markedly inhibits proliferation of HepG2 and SMMC 7721 cells
  • Arrests cell cycle at G0/G1 phase and induces apoptosis
  • Reduces cyclin D1, cyclin E, and CDK2 levels
  • Increases Bax protein and mRNA levels while decreasing Bcl-2
  • Inhibits surgery-induced neuroinflammation and learning impairment in rats
Catalog No. 50-004-07153 Supplier Medchemexpress LLC Supplier No. HYB0402100G
Amantadine is an orally active and potent antiviral agent effective against influenza A viruses and coronaviruses. It inhibits various ion channels, including NMDA and M2, and shows anti-orthopoxvirus and anticancer activities. This compound is suitable for research applications related to Parkinson's disease, postoperative cognitive dysfunction, and COVID-19.
  • Inhibits influenza A viruses and coronavirus ion channels
  • Demonstrates anti-orthopoxvirus and anticancer activity
  • Suitable for research on Parkinson's disease and COVID-19
  • Inhibits SARS-CoV-2 replication in Vero E6 cells
  • Markedly inhibits proliferation of HepG2 and SMMC 7721 cells
  • Arrests cell cycle at G0/G1 phase and induces apoptosis
  • Reduces cyclin D1, cyclin E, and CDK2 levels
  • Increases Bax protein and mRNA levels while decreasing Bcl-2
  • Inhibits surgery-induced neuroinflammation and learning impairment in rats