Learn More
Medchemexpress LLC Amantadine | 768-94-5 | 99.9% | 500 MG
SDP
Catalog No. 5000406703
Encompass_Preferred
Amantadine is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits several ion channels, including NMDA and M2, as well as Coronavirus ion channels. Amantadine also exhibits anti-orthopoxvirus and anticancer activity and can be utilized in research for Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
Catalog No. 50-004-06703 Supplier Medchemexpress LLC Supplier No. HYB0402500MG
Amantadine is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits several ion channels, including NMDA and M2, as well as Coronavirus ion channels. Amantadine also exhibits anti-orthopoxvirus and anticancer activity and can be utilized in research for Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.