Learn More
Medchemexpress LLC Amantadine | 768-94-5 | MFCD00020782 | ≥98% | 250 MG
SDP
Catalog No. 5000406491
Encompass_Preferred
Amantadine hydrochloride is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits several ion channels, including NMDA and M2, and also coronavirus ion channels. Additionally, it possesses anti-orthopoxvirus and anticancer activity. It can be used in research related to Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Orally active and potent antiviral agent
  • Effective against influenza A viruses
  • Inhibits NMDA and M2 ion channels
  • Inhibits coronavirus ion channels
  • Possesses anti-orthopoxvirus activity
  • Possesses anticancer activity
  • Used in Parkinson's disease research
  • Used in postoperative cognitive dysfunction (POCD) research
  • Used in COVID-19 research
Catalog No. 50-004-06491 Supplier Medchemexpress LLC Supplier No. 250MGHYB0402AR
Amantadine hydrochloride is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits several ion channels, including NMDA and M2, and also coronavirus ion channels. Additionally, it possesses anti-orthopoxvirus and anticancer activity. It can be used in research related to Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Orally active and potent antiviral agent
  • Effective against influenza A viruses
  • Inhibits NMDA and M2 ion channels
  • Inhibits coronavirus ion channels
  • Possesses anti-orthopoxvirus activity
  • Possesses anticancer activity
  • Used in Parkinson's disease research
  • Used in postoperative cognitive dysfunction (POCD) research
  • Used in COVID-19 research