Learn More
Medchemexpress LLC Amantadine-d15 hydrochloride | 33830-10-3 | MFCD01317436 | >95.0% | 202.80 g/mol | C10H3D15ClN | 1 MG
SDP
Catalog No. 5000284717
Encompass_Preferred
Amantadine-d15 hydrochloride is the deuterium-labeled hydrochloride salt of amantadine supplied as a stable isotope reference material for analytical and research use. It is intended for use as an internal standard in mass spectrometry, metabolic tracing, and pharmacokinetic studies, and is provided in small quantities suitable for laboratory workflows.
  • Deuterium-labeled hydrochloride salt (d15) for analytical applications.
  • Molecular formula C10H3D15ClN; molecular weight 202.80 g/mol.
  • High purity; reported as >95% (HPLC) for reference-grade material.
  • Used as an internal standard in MS, metabolic tracing, and pharmacokinetic assays.
  • Packaged in small quantities for laboratory use (for example, 1 mg).
Catalog No. 50-002-84717 Supplier Medchemexpress LLC Supplier No. HYW6539051MG
Amantadine-d15 hydrochloride is the deuterium-labeled hydrochloride salt of amantadine supplied as a stable isotope reference material for analytical and research use. It is intended for use as an internal standard in mass spectrometry, metabolic tracing, and pharmacokinetic studies, and is provided in small quantities suitable for laboratory workflows.
  • Deuterium-labeled hydrochloride salt (d15) for analytical applications.
  • Molecular formula C10H3D15ClN; molecular weight 202.80 g/mol.
  • High purity; reported as >95% (HPLC) for reference-grade material.
  • Used as an internal standard in MS, metabolic tracing, and pharmacokinetic assays.
  • Packaged in small quantities for laboratory use (for example, 1 mg).