Learn More
Medchemexpress LLC Amantadine-d6 | 1219805-53-4 | 98.1% | 5 MG
SDP
Catalog No. 5000407018
Encompass_Preferred
Amantadine-d6 is the deuterium labeled Amantadine. Amantadine (1-Adamantanamine) is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits various ion channels, including NMDA and M2, as well as Coronavirus ion channels. It also exhibits anti-orthopoxvirus and anticancer activities. Amantadine-d6 can serve as a tracer and an internal standard for quantitative analysis, with deuteration potentially influencing pharmacokinetic and metabolic profiles of drugs. Applications include research for Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07018 Supplier Medchemexpress LLC Supplier No. HYB0402S15MG
Amantadine-d6 is the deuterium labeled Amantadine. Amantadine (1-Adamantanamine) is an orally active and potent antiviral agent effective against influenza A viruses. It inhibits various ion channels, including NMDA and M2, as well as Coronavirus ion channels. It also exhibits anti-orthopoxvirus and anticancer activities. Amantadine-d6 can serve as a tracer and an internal standard for quantitative analysis, with deuteration potentially influencing pharmacokinetic and metabolic profiles of drugs. Applications include research for Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs