Learn More
Medchemexpress LLC Amantadine-d6 | 1219805-53-4 | MFCD19704894 | 98.1% | 157.29 | C10H11D6N | 10 MG
SDP
Catalog No. 5000298097
Encompass_Preferred
Amantadine-d6 is a deuterium-labeled analogue of amantadine supplied as a stable isotope standard for research and analytical applications, including mass spectrometry, metabolic, and pharmacokinetic studies.
  • Deuterium-labeled (d6) amantadine for stable isotope applications.
  • High chemical and isotopic purity (98.1%).
  • Molecular formula C10H11D6N; molecular weight 157.29.
  • Suitable as an internal standard for LC-MS/MS and other analytical methods.
  • Available in small-mass packaging for analytical workflows (e.g., 10 MG).
Catalog No. 50-002-98097 Supplier Medchemexpress LLC Supplier No. HYB0402S110MG
Amantadine-d6 is a deuterium-labeled analogue of amantadine supplied as a stable isotope standard for research and analytical applications, including mass spectrometry, metabolic, and pharmacokinetic studies.
  • Deuterium-labeled (d6) amantadine for stable isotope applications.
  • High chemical and isotopic purity (98.1%).
  • Molecular formula C10H11D6N; molecular weight 157.29.
  • Suitable as an internal standard for LC-MS/MS and other analytical methods.
  • Available in small-mass packaging for analytical workflows (e.g., 10 MG).