Learn More
Apexbio Technology LLC AMANTADINE-D6-5MG
SDP
Catalog No. 5000569039
Encompass_Preferred
Amantadine-d6 (CAS No 1219805-53-4) is a deuterated form of amantadine commonly used in biomedical research It belongs to the class of adamantane derivatives and is primarily utilized as a tool compound for studying dopaminergic mechanisms and NMDA receptor pathways This compound demonstrates in vitro activity typically in the nanomolar to low micromolar range It is frequently employed in cell models and animal studies for drug screening and mechanistic investigations related to neurological disorders Amantadine-d6 provides insights into viral infections particularly those caused by influenza A viruses It is widely used to investigate resistance mechanisms and drug interactions Experimental concentrations typically depend on the specific research design This compound facilitates the understanding of pharmacokinetics and metabolic pathways through isotopic labeling helping researchers discern subtle biological processes
Catalog No. 50-005-69039 Supplier Apexbio Technology LLC Supplier No. SL10215
Amantadine-d6 (CAS No 1219805-53-4) is a deuterated form of amantadine commonly used in biomedical research It belongs to the class of adamantane derivatives and is primarily utilized as a tool compound for studying dopaminergic mechanisms and NMDA receptor pathways This compound demonstrates in vitro activity typically in the nanomolar to low micromolar range It is frequently employed in cell models and animal studies for drug screening and mechanistic investigations related to neurological disorders Amantadine-d6 provides insights into viral infections particularly those caused by influenza A viruses It is widely used to investigate resistance mechanisms and drug interactions Experimental concentrations typically depend on the specific research design This compound facilitates the understanding of pharmacokinetics and metabolic pathways through isotopic labeling helping researchers discern subtle biological processes